Skip to Content

A3724

Acetyl-β-cyclodextrin

Monoacetyl-β-cyclodextrin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

form

powder

extent of labeling

8 DS (degree of substitution)

color

white

storage temp.

2-8°C

SMILES string

CC(=O)OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C44H72O36/c1-9(51)66-8-16-37-23(58)30(65)44(73-16)79-36-15(7-50)71-42(28(63)21(36)56)77-34-13(5-48)69-40(26(61)19(34)54)75-32-11(3-46)67-38(24(59)17(32)52)74-31-10(2-45)68-39(25(60)18(31)53)76-33-12(4-47)70-41(27(62)20(33)55)78-35-14(6-49)72-43(80-37)29(64)22(35)57/h10-50,52-65H,2-8H2,1H3/t10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-/m1/s1

InChI key

HECSPEHQYGRENG-LZITWAFGSA-N

Application

Cyclodextrins (CD) are cyclic oligosaccharides composed of D-glucose units connected via α(1,4) glycosidic bonds. Common cyclodextrins contain six (α-CD); seven (β-CD) or eight (gamma-CD) D-glucose units. Cyclodextrins are sometimes derivitized through esterification at positions two, three and/or six.
Acetyl-β-cyclodextrin is used in the development of recognition and separation technologies to resolve enantiomers based on chiral properties and size. Acetyl-β-cyclodextrin is used in chiral selection and separation by capillary electrophoresis and liquid chromatography.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library