Skip to Content
Merck
All Photos(2)

Documents

A3547

Sigma-Aldrich

Achromopeptidase from bacteria

lyophilized powder, ≥1,000 units/mg solid

Synonym(s):

ACP, Lysyl endopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (LYSOBACTER SP. BACTERIA M497-1)

Quality Level

form

lyophilized powder

specific activity

≥1,000 units/mg solid

mol wt

27 kDa

technique(s)

cell based assay: suitable

suitability

suitable for cell lysis

application(s)

diagnostic assay manufacturing

shipped in

dry ice

storage temp.

−20°C

Application

Achromopeptidase is useful for lysis of Gram-positive bacteria that are resistant to lysozyme. Achromopeptidase, from Sigma, has been used for the isolation and molecular characterization of skin and soft-tissue bacterial infections.

Biochem/physiol Actions

Achromopeptidase is a lysyl endopeptidase that has a molecular weight of approximately 27 kDa. It works best between pH 8.5 -9. Approximately 500-1,500 μn/ml achromopetidase can be used to lyse cells at a density of OD600 = 0.6 over 2 hours at 37 °C.
Achromopeptidase is a lysyl endopeptidase with a MW of ~27 kDa. It is useful for lysis of Gram-positive bacteria that are resistant to lysozyme.
pH Optimum for activity: pH 8.5 - 9
Approximately 500-1,500 un/ml achromopetidase can be used to lyse cells at a density of OD600=0.6 over 2 hours at 37 °C.

Unit Definition

One unit will lyse 0.4 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 8.0 at 37 °C.

Physical form

crude powder

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Disposable platform for bacterial lysis and nucleic acid amplification based on a single USB-powered printed circuit board.
Shah, et al.
PLoS ONE, 18, e0284424-e0284424 (2023)
Georgios Chondrogiannis et al.
Scientific reports, 11(1), 6140-6140 (2021-03-19)
Enzymes are the cornerstone of modern biotechnology. Achromopeptidase (ACP) is a well-known enzyme that hydrolyzes a number of proteins, notably proteins on the surface of Gram-positive bacteria. It is therefore used for sample preparation in nucleic acid tests. However, ACP
Yuanyuan Hu et al.
World journal of microbiology & biotechnology, 32(1), 1-1 (2015-11-26)
Extracting DNA from Staphylococcus aureus cells is important for detecting MRSA by PCR. However, S. aureus cells are known to be difficult to disrupt due to their compact cell walls. Here, we systematically studied the efficiency of a highly active
Chelsey A Smith et al.
Lab on a chip, 23(3), 451-465 (2022-12-24)
Cervical cancer is a leading cause of cancer death for women in low-resource settings. The World Health Organization recommends that cervical cancer screening programs incorporate HPV DNA testing, but available tests are expensive, require laboratory infrastructure, and cannot be performed
J R Buser et al.
Analytical methods : advancing methods and applications, 8(14), 2880-2886 (2016-04-14)
Sample preparation, including bacterial lysis, remains a hurdle in the realization of complete point-of-care tests for many pathogens. Here, we developed a sample preparation methodology for enzymatic lysis and sample heating for low-resource, point-of-care applications. We show an instrument-free chemical

Protocols

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

To measure achromopeptidase activity, this procedure uses bacterial cells and a turbidimetric rate assay. Turbidity is measured at 600 nm using a spectrophotometer.

Related Content

Find cell lysis and protein extraction reagents like cell lysis buffers, protein extraction kits, extraction enzymes, and other lysis enhancers for isolating and purifying proteins.

Find cell lysis and protein extraction reagents like cell lysis buffers, protein extraction kits, extraction enzymes, and other lysis enhancers for isolating and purifying proteins.

Find cell lysis and protein extraction reagents like cell lysis buffers, protein extraction kits, extraction enzymes, and other lysis enhancers for isolating and purifying proteins.

Find cell lysis and protein extraction reagents like cell lysis buffers, protein extraction kits, extraction enzymes, and other lysis enhancers for isolating and purifying proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service