Skip to Content

A3106

Apolipoprotein C-III from human plasma

buffered aqueous solution

Synonym(s):

Apo C-III

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.61
MDL number:
Form:
buffered aqueous solution
Assay:
≥95% (SDS-GE)
Biological source:
human plasma
Mol wt:
8.75 kDa
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human plasma

Quality Segment

assay

≥95% (SDS-GE)

form

buffered aqueous solution

mol wt

8.75 kDa

technique(s)

electrophoresis: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APOC3(345)

Biochem/physiol Actions

Apolipoprotein C-III (apoC-III) is central in regulating the metabolism of triglyceride-rich lipoproteins. It is an inhibitor of lipoprotein lipase (LPL).
Apolipoprotein C-III (apoC-III) is central in regulating the metabolism of triglyceride-rich lipoproteins. It is an inhibitor of lipoprotein lipase (LPL). Plasma levels of apoC-III are somewhat correlated to body mass index (BMI) and insulin sensitivity, and strongly correlated to levels of VLDL triglycerides. Reduced levels of apoC-III lead to higher fatty acid uptake of plasma triglycerides into adipose tissue.

Physical form

Solution in 10 mM ammonium bicarbonate.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library