Skip to Content
Merck

A3041

β-Alanine t-butyl ester hydrochloride

≥98% (TLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H15NO2 · HCl
CAS Number:
Molecular Weight:
181.66
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

β-Alanine t-butyl ester hydrochloride,

assay

≥98% (TLC)

Quality Segment

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

Cl.CC(C)(C)OC(=O)CCN

InChI

1S/C7H15NO2.ClH/c1-7(2,3)10-6(9)4-5-8;/h4-5,8H2,1-3H3;1H

InChI key

DOMTZTVJNZKUNX-UHFFFAOYSA-N

Biochem/physiol Actions

β-Alanine t-butyl ester is a C-terminal blocked derivative of β-alanine.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
A3041-5G04061832421056