A2973
ART® self-sealing barrier pipette tips
ART 100E, volume range 1-100 μL, sterile
Synonym(s):
pipet tips, pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
volume:
1—100 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
RNase free
barrier
packaging:
case of 5 packs
pack of 960 ea (Racked)
pack of 960 ea (Racked)
manufacturer/tradename:
Molecular BioProducts 2065E
Recommended Products
material
colorless
sterility
sterile
feature
DNase free
RNase free
barrier
packaging
case of 5 packs
pack of 960 ea (Racked)
manufacturer/tradename
Molecular BioProducts 2065E
technique(s)
PCR: suitable
maximum volume
100 μL
volume range
1-100 μL
volume
1—100 μL
Looking for similar products? Visit Product Comparison Guide
Application
ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).
Packaging
960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)
800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA
4800EA = 5 packs of 960 tips (50 trays of 96 tips)
800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA
Compatibility
Compatibility
Biohit Electronic 10-100 μl
Biohit Variable 5-50 μl
Corning 4962 10-100 μl
Eppendorf 4810 10-100 μl
Eppendorf Response 4850 5-100 μl
Eppendorf Research Pro 4860 5-100 μl/50-1000 μl
Eppendorf Titerman 4908 30-300 μl
Hamilton/SoftGrip Precision 100 μl
Mettler-Toledo VoluMate 10-100 μl
MLA Digital Pipette 10-50 μl
Nichiryo BenchMate 5-100 μl
Pipetman P-100 10-100 μl
Rainin EDP 2 10-100 μl
Rainin EDP Plus 10-100 μl
Rainin Pipet Plus 10-100 μl
Rainin Pipetman 10-100 μl
Transferpette 5-50 μl
Transferpette Multi-Channel 10-100 μl
Biohit Electronic 10-100 μl
Biohit Variable 5-50 μl
Corning 4962 10-100 μl
Eppendorf 4810 10-100 μl
Eppendorf Response 4850 5-100 μl
Eppendorf Research Pro 4860 5-100 μl/50-1000 μl
Eppendorf Titerman 4908 30-300 μl
Hamilton/SoftGrip Precision 100 μl
Mettler-Toledo VoluMate 10-100 μl
MLA Digital Pipette 10-50 μl
Nichiryo BenchMate 5-100 μl
Pipetman P-100 10-100 μl
Rainin EDP 2 10-100 μl
Rainin EDP Plus 10-100 μl
Rainin Pipet Plus 10-100 μl
Rainin Pipetman 10-100 μl
Transferpette 5-50 μl
Transferpette Multi-Channel 10-100 μl
Other Notes
Browse all pipette tips
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)
Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service