Skip to Content
Merck
All Photos(2)

Key Documents

A2556

Sigma-Aldrich

Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse

clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution

Synonym(s):

Monoclonal Anti-Rabbit IgG (γ-chain specific)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

RG-96, monoclonal

form

buffered aqueous glycerol solution

species reactivity

rabbit

should not react with

goat, feline, pig, guinea pig, rat, bovine, canine, human, horse, sheep, chicken

technique(s)

direct ELISA: 1:50,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100
western blot: 1:200,000-1:400,000 using total cell extract of HeLa cells

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Rabbit IgG (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. Immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It is a glycoprotein and mainly helps in immune defense. IgG is a major class of immunoglobulin. Rabbit IgG is further divided into five classes- IgM, IgG, IgA, IgD and IgE.
The product recognizes an epitope located on the γ (heavy)-chain of rabbit IgG. In immunoblotting, the antibody recognizes both native and denatured forms of rabbit IgG. In ELISA, the antibody is specific for rabbit IgG, and shows no cross-reactivity with rabbit IgA and IgM or human IgG, IgA, and IgM. No cross reaction is observed with IgG from the following species: bovine, cat, chicken, dog, goat, guinea pig, horse, pig, rat, or sheep.

Application

Monoclonal Anti-Rabbit IgG (γ-chain specific)Alkaline Phosphatase antibody produced in mouse has been used in enzyme-linked immunosorbent assay (ELISA) and immunoblotting.

Biochem/physiol Actions

Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. It helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of the megakaryocytic glycoprotein IX promoter by the oncogenic Ets transcription factor Fli-1
Bastian LS, et al.
Blood, 93(8), 2637-2644 (1999)
Heidi C Schmid et al.
Biological chemistry, 383(11), 1769-1778 (2003-01-18)
The reaction of recombinant chlorophyll synthase from Avena sativa, expressed in Escherichia coli, was investigated. To verify the identity of the recombinant and native enzymes, reaction rates were determined for both enzyme preparations with several chlorophyllide analogs. The rates of
The glycosylation of human serum IgD and IgE and the accessibility of identified oligomannose structures for interaction with mannan-binding lectin
Arnold JN, et al.
Journal of Immunology, 173(11), 6831-6840 (2004)
Wen-Hao Tang et al.
Molecular medicine reports, 17(2), 2257-2262 (2017-12-06)
The aim of the present study was to explore the underlying mechanism and diagnostic potential of Ran‑binding protein M (RanBPM) in human spermatogenesis and oogenesis. RanBPM expression in human testis and ovaries was analysed using polymerase chain reaction (PCR) and
Nicholas Svitek et al.
Veterinary research, 45, 50-50 (2014-04-30)
Peptide-major histocompatibility complex (p-MHC) class I tetramer complexes have facilitated the early detection and functional characterisation of epitope specific CD8+ cytotoxic T lymphocytes (CTL). Here, we report on the generation of seven recombinant bovine leukocyte antigens (BoLA) and recombinant bovine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service