Skip to Content
Merck
All Photos(7)

Key Documents

A2103

Sigma-Aldrich

Anti-γ-Actin (ACTG1) Antibody

enhanced validation

rabbit polyclonal

Synonym(s):

Actin Detection Antibody, Rabbit Anti-Actin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-Actin, N-terminal antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 42 kDa

species reactivity

frog, rat, mouse, chicken, human

packaging

antibody small pack of 25 μL

enhanced validation

independent
Learn more about Antibody Enhanced Validation

concentration

~0.5 mg/mL

technique(s)

immunocytochemistry: 1-2 μg/mL using cultured chicken fibroblasts
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 2-4 μg/mL using sections of human appendix, mouse heart, and frog skeletal muscle
indirect immunofluorescence: suitable
western blot: 0.5-1 μg/mL using whole extract of the human epitheloid carcinoma HeLa cell line.
western blot: 2-4 ng/mL using whole extract of rat skeletal muscle

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Actin is a cytoskeletal protein that regulates cell motility, secretion, phagocytosis and cytokinesis. The NH2-terminal of actin may function as an antigen. This terminal may also modulate actin interactions and may associate with proteins such as myosin.
Actin is one of the most conserved eukaryotic proteins, which is expressed in mammals and birds as at least six major different isoforms. Four of them represent the differentiation markers of muscle tissues and two are found practically in all non-muscle cells. Actin isoforms show >90% overall sequence homology, but only 50-60% homology in their 18 N-terminal residues. Actin is present in cells both as a globular monomer (G-actin) and as a polymer in filamentous actin (F-actin) that participates in the formation of a variety of stable and labile structures. The presence of actin in cell nuclei and cell membranes has been reported.

Immunogen

synthetic actin N-terminal nonapeptide conjugated to KLH.

Application

Anti-Actin, N-terminal antibody produced in rabbit has been used in
  • immunoblotting
  • quantitative chromatin immunoprecipitation (qChIP)

Protein lysates generated from embryonic mouse limb tissue was analyzed by western blot using actin as the loading control. Actin was detected using rabbit anti-actin at 1:8000. Actin detected by a rabbit anti-actin antibody was used as a loading control for MCF7 cell lysates . Actin was detected at 42 kDa.
Rabbit anti-actin, N-terminal antibody can be used for western blotting assays. The antibody can also be used for immunocytochemistry at 1-2μg/mL using cultured chicken fibroblasts. Furthermore, the antibody is suitable for use in immunohistochemistry at a concentration of 2-4μg/mL using sections of human appendix, mouse heart, and frog skeletal muscle.

Biochem/physiol Actions

Actin and myosin are constituents of many cell types and are involved in a myriad of cellular processes including maintenance of cell shape, locomotion, secretion, cytoplasmic streaming, phagocytosis, and cytokinesis. The N-terminal domain of actin appears to be a major antigenic region of the molecule. Different isoforms can coexist in the same cell but are differentially regulated, and in most cases cannot substitute for each other functionally.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Other Notes

To view an Actin antibody selection guide, please visit www.sigmaaldrich.com/actin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monoclonal antibodies against muscle actin isoforms: epitope identification and analysis of isoform expression by immunoblot and immunostaining in normal and regenerating skeletal muscle
Chaponnier C and Gabbiani G
F1000Research, 5 (2016)
Luciferase expression and bioluminescence does not affect tumor cell growth in vitro or in vivo
Tiffen JC, et al.
Molecular Cancer, 9(1), 299-299 (2010)
Nuclear actin polymerization is required for transcriptional reprogramming of Oct4 by oocytes
Miyamoto K, et al.
Genes & Development, 25(9), 946-958 (2011)
Kale S Bongers et al.
American journal of physiology. Endocrinology and metabolism, 305(7), E907-E915 (2013-08-15)
Skeletal muscle denervation causes muscle atrophy via complex molecular mechanisms that are not well understood. To better understand these mechanisms, we investigated how muscle denervation increases growth arrest and DNA damage-inducible 45α (Gadd45a) mRNA in skeletal muscle. Previous studies established
Actin, a central player in cell shape and movement
Pollard TD and Cooper JA
Science, 326(5957), 1208-1212 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service