Skip to Content

A1520

α-Amylase Inhibitor

≥200 inhibitor U/mg protein (using porcine pancreatic α-amylase), ≥1000 inhibitor U/mg protein (using human salivary α-amylase), lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

α-Amylase Inhibitor from Triticum aestivum (wheat seed), Type I, lyophilized powder

biological source

Triticum estivum

Quality Segment

type

Type I

form

lyophilized powder

specific activity

≥1000 inhibitor U/mg protein (using human salivary α-amylase), ≥200 inhibitor U/mg protein (using porcine pancreatic α-amylase)

composition

Protein, 35-65% biuret

input

plant, fungi and insect sample(s)

storage temp.

−20°C

General description

α-Amylases are endoglycosidases, that are classified into glycosyl hydrolase family 13 (1-3).

Application

α-Amylase Inhibitor from Triticum aestivum (wheat seed) has been used to determine its effects on salivary gland (SG) amylase activity. It has also been used to inhibit the activity from the extract from Bemisia tabaci.

Biochem/physiol Actions

Competitive inhibitor of human salivary α-amylase. KI = 2.9 nM, compared to a KM of 5.9 mM (calculated per mole of α-1,4-linked maltose residues).
α-Amylases can hydrolyze α-(1,4)-D-glycosidic linkages. It participates in the breakdown of starch and glycogen. Inhibiting this enzyme can be used as a strategy to treat dental caries, diabetes, obesity and periodontal diseases.

Physical form

Lyophilized powder containing buffer salts as sodium phosphate.

Other Notes

One unit will reduce the activity of two units of α-amylase (A0521) by 50% after pre-incubation at 25 °C.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library