Skip to Content
Merck
All Photos(1)

Documents

A1054

Sigma-Aldrich

Anti-Digoxin–Alkaline Phosphatase antibody, Mouse monoclonal

clone DI-22, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Digoxin, Monoclonal Anti-Digoxin–Alkaline Phosphatase antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

recombinant

expressed in mouse cell line

conjugate

alkaline phosphatase conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

DI-22, monoclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:10,000 using using 20 μg/ml of purified digoxin-BSA for coating and pNPP substrate
direct ELISA: 1:6,000 using using 10 μg/ml of digoxigenin-transferrin for coating and pNPP substrate
dot blot: 1:20,000 using digoxigenin-labeled biomolecules (chemiluminescent)

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Digoxin is a cardiac glycoside that is used to treat chronic heart failure.Monoclonal Anti-Digoxin-Alkaline Phosphatase antibody can be used to detect digoxin-labeled compounds such as oligonucleotides, antibodies and peptides. The antibody is specific for digoxin and digoxin-labeled compounds. The product shows strong cross-reactivity with digoxigenin.

Specificity

Antibody is specific for digoxin and shows a high affinity for digoxigenin.

Immunogen

Digoxin-KLH.

Application

Monoclonal Anti-Digoxin-Alkaline Phosphatase antibody is suitable for use in direct ELISA (1:10000-1:6000) and dot blot (at 1:20,000 dilutions using digoxigenin-labeled biomolecules). It has also been used for enzyme immunoassay (EIA).
Nested PCR products were hybridized with 5′-end digoxigenin-labeled oligonucleotide probe. ELISA assays were performed to detect the probe by adding monoclonal anti-digoxin antibody alkaline phosphatase (DI-22) diluted 1:1000 in 1 M phosphate-buffered saline, 1% bovine serum albumin, and 0.1% Tween 20 (Ph 7.4). Detection was performed using Sigma Fass-Pnpp substrate tablets.

Physical form

Solution in 0.05 M Tris buffer, pH 8.0 containing 1% BSA, 50% glycerol, 1 mM MgCl2 and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J N Mbatha et al.
BMC research notes, 10(1), 702-702 (2017-12-07)
Cervical cancer is a major problem in women and it is important to find a suitable and acceptable screening method, especially among young in low-resource areas for future human papillomavirus (HPV) vaccine follow-up investigations. The study sought to test the
Joyce N Mbatha et al.
Infectious diseases (London, England), 49(8), 601-608 (2017-04-14)
High-risk human papillomavirus (hr-HPV) infections and low-grade squamous intraepithelial lesions occur frequently in young women. The available vaccines cover up to seven hr-HPV genotypes (HPV16, HPV18, HPV31, HPV33, HPV45, HPV52 and HPV58) and two low-risk HPV types (HPV6 and HPV11).
The New England journal of medicine, 336(8), 525-533 (1997-02-20)
The role of cardiac glycosides in treating patients with chronic heart failure and normal sinus rhythm remains controversial. We studied the effect of digoxin on mortality and hospitalization in a randomized, double-blind clinical trial. In the main trial, patients with
Self-sampling for human papillomavirus testing among rural young women of KwaZulu-Natal, South Africa
Mbatha JN, et al.
BMC Research Notes, 10(1), 1-8 (2017)
Liang Wang et al.
Theranostics, 11(2), 925-940 (2021-01-05)
Background: Asparaginase (ASP) is the cornerstone drug in the treatment of extranodal NK/T-cell lymphoma (ENKTCL), and the mechanisms of resistance to ASP remain largely unknown. Long non-coding RNAs play important roles in chemotherapy resistance in various cancers. However, the expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service