Skip to Content
Merck
All Photos(1)

Key Documents

94747

Sigma-Aldrich

Vancomycin hydrochloride from Streptomyces orientalis

≥85% (Vancomycin B)

Synonym(s):

Vancomycin HCL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H75Cl2N9O24 · HCl
CAS Number:
Molecular Weight:
1485.71
Beilstein:
3704657
MDL number:
UNSPSC Code:
51282703
NACRES:
NA.85

biological source

Streptomyces orientalis

Quality Level

Assay

≥85% (Vancomycin B)

form

powder

color

, off-white to brown or White to orange-brown

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

Cl[H].CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]1O[C@H]2C[C@](C)(N)[C@@H](O)[C@@H](C)O2.CN[C@H](CC(C)C)C(=O)NC3[C@H](O)c4ccc(Oc5cc6Oc7ccc(cc7Cl)[C@@H](O)[C@H]8NC(=O)[C@H](NC(=O)[C@H](NC(=O)[C@H](CC(N)=O)NC3=O)c(c5)c6)c9ccc(O)c(c9)-c%10c(O)cc(O)cc%10[C@@H](NC8=O)C(O)=O)c(Cl)c4

InChI

1S/C66H75Cl2N9O24.ClH/c1-23(2)12-34(71-5)58(88)76-49-51(83)26-7-10-38(32(67)14-26)97-40-16-28-17-41(55(40)101-65-56(54(86)53(85)42(22-78)99-65)100-44-21-66(4,70)57(87)24(3)96-44)98-39-11-8-27(15-33(39)68)52(84)50-63(93)75-48(64(94)95)31-18-29(79)19-37(81)45(31)30-13-25(6-9-36(30)80)46(60(90)77-50)74-61(91)47(28)73-59(89)35(20-43(69)82)72-62(49)92;/h6-11,13-19,23-24,34-35,42,44,46-54,56-57,65,71,78-81,83-87H,12,20-22,70H2,1-5H3,(H2,69,82)(H,72,92)(H,73,89)(H,74,91)(H,75,93)(H,76,88)(H,77,90)(H,94,95);1H/t24-,34+,35-,42+,44-,46+,47+,48-,49+,50-,51+,52+,53+,54-,56+,57+,65-,66-;/m0./s1

InChI key

LCTORFDMHNKUSG-XTTLPDOESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: glycopeptide

Application

Vancomycin has been used to study transcriptional induction of the penicillin-binding protein 2 gene in Staphylococcus aureus, the correlation of vancomycin and daptomycin susceptibility in Staphylococcus aureus, and the evaluation of telavancin activity in Staphylococcus aureus.

Biochem/physiol Actions

Mode of action: Inhibits incorporation of terminal D-alanyl-D-alanine moieties of the NAM/NAG-peptides.

Antimicrobial Spectrum: Active against Gram-positive bacteria

Packaging

250mg, 1g, 5g

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helio S Sader et al.
The Journal of antimicrobial chemotherapy, 64(5), 1024-1028 (2009-09-12)
The bactericidal activities of vancomycin and daptomycin were evaluated in a large collection of methicillin-resistant Staphylococcus aureus (MRSA) bacteraemia strains from nine major USA medical centres. To evaluate the occurrence of heterogeneous vancomycin-intermediate S. aureus (hVISA) among MRSA strains tolerant
Gabriel A Russell et al.
STAR protocols, 2(4), 100946-100946 (2021-11-27)
The microbes indigenous to helminth species are a major obstacle to deciphering host-parasite interactions. Repurposing a system of reversible bacterial colonization, we have generated germ-free Heligomosomoides polygyrus bakeri (Hpb) larvae that maintain the sterility of axenic mice upon infection. This protocol
Warren E Rose et al.
The Journal of antimicrobial chemotherapy, 59(6), 1190-1193 (2007-04-17)
Recently, an association between the accessory gene regulator (agr) in Staphylococcus aureus and the development of vancomycin resistance secondary to suboptimal exposure has been demonstrated. We investigated the relationship of vancomycin with and without gentamicin or rifampicin, and daptomycin in
Susan Boyle-Vavra et al.
Antimicrobial agents and chemotherapy, 47(3), 1028-1036 (2003-02-27)
We found an increased abundance of pbpB-specific transcripts in vancomycin intermediate-resistant Staphylococcus aureus (VISA) isolates compared with that found in paired, genetically identical, susceptible isolates. This difference in expression cannot be explained by differences in the pbpB promoter sequence. Since
Kerri A Miller et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(13), 5955-5960 (2019-03-10)
Copper is a critical enzyme cofactor in the body but also a potent cellular toxin when intracellularly unbound. Thus, there is a delicate balance of intracellular copper, maintained by a series of complex interactions between the metal and specific copper

Articles

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service