Skip to Content
Merck
All Photos(1)

Key Documents

94577

Sigma-Aldrich

DL-α-Hydroxyglutaric acid disodium salt

≥95.0% (GC)

Synonym(s):

(RS)-2-Hydroxypentanedioic acid disodium salt, Disodium 2-hydroxyglutarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H6O5Na2
CAS Number:
Molecular Weight:
192.08
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.28

Assay

≥95.0% (GC)

form

powder or crystals

impurities

≤6.0% water

storage temp.

2-8°C
2-8°C

SMILES string

[Na+].[Na+].OC(CCC([O-])=O)C([O-])=O

InChI

1S/C5H8O5.2Na/c6-3(5(9)10)1-2-4(7)8;;/h3,6H,1-2H2,(H,7,8)(H,9,10);;/q;2*+1/p-2

InChI key

DZHFTEDSQFPDPP-UHFFFAOYSA-L

Biochem/physiol Actions

D-(or R-) enantiomer of α-hydroxyglutaric acid is a cancer related biomarker of diagnostic value, whereas the L-(or S-) enantiomer of α-hydroxyglutaric acid is a biomarker for certain neurometabolic, inherited disorder diseases. It has no known physiological function in eukaryotes. It is toxic to the brain and increases the susceptibility to develop brain tumors.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cancer: A metabolic metamorphosis.
Abigail S Krall et al.
Nature, 496(7443), 38-40 (2013-04-05)
Jelena Lazovic et al.
Neuro-oncology, 14(12), 1465-1472 (2012-10-24)
The arginine 132 (R132) mutation of isocitrate dehydrogenase -1 (IDH1(R132)) results in production of 2-hydroxyglutarate (2-HG) and is associated with a better prognosis compared with wild-type (WT) in glioma patients. The majority of lower-grade gliomas express IDH1(R132), whereas this mutation
Fang Wang et al.
Science (New York, N.Y.), 340(6132), 622-626 (2013-04-06)
A number of human cancers harbor somatic point mutations in the genes encoding isocitrate dehydrogenases 1 and 2 (IDH1 and IDH2). These mutations alter residues in the enzyme active sites and confer a gain-of-function in cancer cells, resulting in the
Cancer-associated IDH mutations: biomarker and therapeutic opportunities
K E Yen
Oncogene (2010)
Cancer-associated IDH1 mutations produce 2-hydroxyglutarate
Lenny Dang
Nature (2009)

Protocols

We describe here a rapid and sensitive method to separate and measure D-2-OHG and L-2-OHG enantiomers using high-resolution mass spectrometry (HRMS) detection.

Chromatograms

application for HPLCapplication for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service