Skip to Content
Merck

934B

PBS IHC Wash Buffer + TWEEN® 20

20X PBS, 1% Tween® 20

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

[For In Vitro Diagnostic Use in Select Regions]

form

buffered aqueous solution

packaging

pkg of 1 L 20x (934B-09), pkg of 200 mL 20x (934B-06)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry: suitable

shipped in

ambient

General description

Phosphate Buffered Immunohistochemistry wash buffer plus TWEEN 20 is a 20X concentrated solution that is employed to rinse reagents off slides and to provide a medium for short-term storage of immunohistochemistry specimens between application of reagents.

Physical form

Solution containing PBS buffer and TWEEN 20

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library