Skip to Content
Merck

92243

Cholesteryl N-(2-dimethylaminoethyl)carbamate

≥98% (TLC)

Synonym(s):

3β-{N-[2-(Dimethylamino)ethyl]carbamoyl}cholesterol, DC-Chol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H56N2O2
CAS Number:
Molecular Weight:
500.80
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98% (TLC)

form

powder or crystals

functional group

ester

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4CC(CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)NCCN(C)C

InChI

1S/C32H56N2O2/c1-22(2)9-8-10-23(3)27-13-14-28-26-12-11-24-21-25(36-30(35)33-19-20-34(6)7)15-17-31(24,4)29(26)16-18-32(27,28)5/h11,22-23,25-29H,8-10,12-21H2,1-7H3,(H,33,35)/t23-,25?,26+,27-,28+,29+,31+,32-/m1/s1

InChI key

HIHOWBSBBDRPDW-MTIZRNOUSA-N

Application


  • Role of interleukin-6 in antigen-specific mucosal immunoglobulin A induction by cationic liposomes.: This study investigates the use of cholesteryl N-(2-dimethylaminoethyl)carbamate in cationic liposomes for enhancing mucosal immunoglobulin A production, highlighting its potential in vaccine development (Tada et al., 2021).

  • Nasal vaccination with pneumococcal surface protein A in combination with cationic liposomes consisting of DOTAP and DC-chol confers antigen-mediated protective immunity against Streptococcus pneumoniae infections in mice.: This research demonstrates the efficacy of cholesteryl N-(2-dimethylaminoethyl)carbamate in nasal vaccines, providing a promising strategy for respiratory infection prevention (Tada et al., 2018).


Biochem/physiol Actions

Cationic liposome, investigated in cancer gene therapy, as vaccine delivery system/adjuvant.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Malou Henriksen-Lacey et al.
Molecular pharmaceutics, 8(1), 153-161 (2010-12-02)
The immunostimulatory capacities of cationic liposomes are well-documented and are attributed both to inherent immunogenicity of the cationic lipid and more physical capacities such as the formation of antigen depots and antigen delivery. Very few studies have however been conducted
Gene therapy using DC-Chol liposomes.
Goyal, K. and Huang, L.
Journal of liposome research, 5, 49-60 (1995)
Francesco Cardarelli et al.
Molecular pharmaceutics, 9(2), 334-340 (2011-12-27)
Here we investigate the cellular uptake mechanism and final intracellular fate of two cationic liposome formulations characterized by similar physicochemical properties but very different lipid composition and efficiency for intracellular delivery of DNA. The first formulation is made of cationic



Global Trade Item Number

SKUGTIN
92243-100MG04061833073742
92243-500MG04061833073759