Skip to Content

90678

Lipase, immoblilized on Immobead 150 from Pseudomonas fluorescens

≥600 U/g

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.54
UNSPSC Code:
12352204
Specific activity:
≥600 U/g
Biological source:
Pseudomonas fluorescens
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Pseudomonas fluorescens

Quality Segment

form

powder

specific activity

≥600 U/g

storage temp.

2-8°C

Application

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Biochem/physiol Actions

Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Other Notes

1 U corresponds to the amount of enzyme which liberates 1 μmol butyric acid per minute at pH 7.5 and 40°C (glyceryl tributyrate, Cat. No. 91010, as substrate)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library