Skip to Content
Merck

861

Naphthol AS-BI Alkaline Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352106
NACRES:
NA.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

4 mg/mL

pH

9.5

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Application

Kit component for Sigma Alkaline Phosphatase Procedure, kits 86C and 86R

Other Notes

Napthol AS-BI Alkaline Solution, 4 mg/mL in AMPD buffer, 2 mol/L, pH 9.5


pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



B Claire McDyre et al.
Toxicologic pathology, 46(2), 202-223 (2018-01-31)
The past decade has seen an increase in the development and clinical use of biomarkers associated with histological features of liver disease. Here, we conduct a comparative histological and global proteomics analysis to identify coregulated modules of proteins in the
Shervin Bahrami et al.
Clinical immunology (Orlando, Fla.), 191, 37-43 (2018-03-24)
Retroviruses including Human Endogenous Retroviruses (HERVs), contain a conserved region with highly immunomodulatory functions in the transmembrane proteins in envelope gene (env) named immunosuppressive domain (ISU). In this report, we demonstrate that Env59-GP3 peptide holds therapeutic potential in a mouse



Global Trade Item Number

SKUGTIN
ABE21704053252910777
SAB4504404-100UG04061832525662
F9176-1VL04061837913693
861-10ML04061833039984
NIST86104061834187059
PL861-CGA-1EA04061832876290
PL861-CG-1EA04061832876252