Skip to Content
Merck
All Photos(1)

Key Documents

82415

Sigma-Aldrich

Prunetin

≥98.0% (TLC)

Synonym(s):

4′,5-Dihydroxy-7-methoxyisoflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
Beilstein:
292155
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98.0% (TLC)

form

powder

SMILES string

COc1cc(O)c2C(=O)C(=COc2c1)c3ccc(O)cc3

InChI

1S/C16H12O5/c1-20-11-6-13(18)15-14(7-11)21-8-12(16(15)19)9-2-4-10(17)5-3-9/h2-8,17-18H,1H3

InChI key

KQMVAGISDHMXJJ-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibitor of both cytoplasmic and mitochondrial human liver aldehyde dehydrogenases, possibly by binding at an allosteric site.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Philippe Antignac et al.
Rapid communications in mass spectrometry : RCM, 17(12), 1256-1264 (2003-06-18)
In an international context of promoting scientific research on food safety, the interest in molecules having potential hormonal disrupting effects is growing. While industrial endocrine disruptors (phthalates, alkylphenols, PCBs, etc.) have been studied for several years, natural compounds like phytoestrogens
M L Shen et al.
Journal of the American Society for Mass Spectrometry, 12(1), 97-104 (2001-01-06)
Aldehyde dehydrogenases (ALDH) are a family of enzymes primarily involved in the oxidation of various aldehydes. Most ALDH enzymes derived from mammalian sources have been shown to exist as homotetramers, consisting of four identical subunits of approximately 54 kDa. The
Nattaya Ngamrojanavanich et al.
Journal of ethnopharmacology, 109(2), 354-358 (2006-09-16)
A carpin (3-hydroxy-9-methoxypterocarpan) (Medicarpin) (1) and four isoflavones, 7-hydroxy-4'-methoxy-isoflavone (Formononetin) (2); 7,4'-dimethoxyisoflavone (3); 5,4'-dihydroxy-7-methoxy-isoflavone (Prunetin) (4) and 7-hydroxy-6,4'-dimethoxyisoflavone (5) were isolated from the tuber roots of Butea superba Roxb. Compounds 2 and 4 showed moderate cytotoxic activity on KB cell
Tae-Gue Ahn et al.
Biochemical pharmacology, 85(10), 1525-1533 (2013-02-27)
Prunetin is an O-methylated isoflavone, which is a type of flavonoid. There are a limited number of reports detailing the biological activities of prunetin. Although an anti-inflammatory effect of prunetin has been reported in vitro, to our knowledge, there have
N S Nagarajan et al.
Natural product research, 20(2), 195-200 (2005-12-02)
Two isoflavonoids isolated from Dalbergia sympathetica were identified as 5,4'-dihydroxy-7-methoxyisoflavone (1) (Prunetin) and Prunetin-4'-O-beta-D-gentiobioside (2) (Dalsympathetin). The natural occurrence of Dalsympathetin is reported for the first time. The position of glycosylation in Dalsympathetin at 4'-position has been confirmed by 2D-NMR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service