Skip to Content
Merck
All Photos(1)

Key Documents

79383

Sigma-Aldrich

Phosphate buffered saline

10X, liquid, pH ~7.4 (25 °C, after tenfold dilution), BioUltra

Synonym(s):

PBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105319
eCl@ss:
32129211
NACRES:
NA.25

Product Name

Phosphate buffered saline, BioUltra, washing buffer for peroxidase conjugates in Western Blotting, 10x concentrate

product line

BioUltra

form

liquid

quality

10x concentrate

impurities

insoluble matter, passes filter test

pH

~7.4 (25 °C, after tenfold dilution)

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01

Looking for similar products? Visit Product Comparison Guide

Application


  • Polyvinylalcohol Composite Filled with Carbon Dots Produced by Laser Ablation in Liquids.: This study explores the synthesis of polyvinyl alcohol (PVA) composites filled with carbon dots (CDs) using laser ablation in liquids (LAL). The composites were characterized and tested for potential biomedical applications, demonstrating enhanced mechanical properties and biocompatibility when evaluated in phosphate-buffered saline (PBS) solution (Cutroneo et al., 2024).

  • Exploring the Interplay between Tribocorrosion and Surface Chemistry of the ASTM F139 Surgical Stainless Steel in Phosphate-Buffered Saline Solution.: This research investigates the tribocorrosion behavior and surface chemistry of ASTM F139 surgical stainless steel in PBS. The study provides insights into the material′s performance and degradation mechanisms under physiological conditions, which is critical for biomedical implant applications (Macedo et al., 2024).

  • Biological Evaluation of Oral Care Products Using 3D Tissue-Engineered In Vitro Models of Plaque-Induced Gingivitis.: This study evaluates the efficacy of various oral care products using 3D tissue-engineered models of plaque-induced gingivitis. PBS was used as a control to simulate physiological conditions during the assessment of the anti-inflammatory and antimicrobial properties of the products (Barker et al., 2024).

  • UCST-Type Thermoresponsive Sol-Gel Transition Triblock Copolymer Containing Zwitterionic Polymer Blocks.: The research focuses on the development of a novel upper critical solution temperature (UCST) thermoresponsive sol-gel transition triblock copolymer. The material′s properties were extensively studied in PBS to evaluate its potential for drug delivery and other biomedical applications (Kawamura et al., 2024).

  • The Tardigrade-Derived Mitochondrial Abundant Heat Soluble Protein Improves Adipose-Derived Stem Cell Survival against Representative Stressors.: This study examines the protective effects of a tardigrade-derived protein on adipose-derived stem cells under stress conditions. The cells′ viability and function were assessed in PBS, providing valuable insights into the protein′s potential for enhancing stem cell therapies (Rolsma et al., 2024).

Caution

precipitate may be redissolved by gently heating

Physical form

100 mM sodium phosphate; 9.0% NaCl; pH 6.8±0.1 (25°C)

Reconstitution

shake well before use

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Ibrahim et al.
Invertebrate neuroscience : IN, 20(4), 16-16 (2020-09-03)
Electron microscopy revealed that glial cells in the posterior sub-esophageal mass of the brain in Sepia officinalis had a well-developed rough endoplasmic reticulum formed by long coverslips with rectilinear or curvilinear arrangements. The coverslips appeared dilated and have a large
Konrad Franke et al.
Translational oncology, 2(4), 310-320 (2009-12-04)
Nodal spread is the single most important prognostic factor of survival in gastric cancer patients. In this study, genes that were upregulated in the lymph node metastases of gastric cancer were identified and may serve as putative novel therapeutic target.
Yong N Li et al.
The Journal of comparative neurology, 516(5), 442-453 (2009-08-06)
Horizontal cells (HCs) are involved in establishing the center-surround receptive field organization of photoreceptor and bipolar cells. In many species, HCs respond differentially to colors and may play a role in color vision. An earlier study from our laboratory suggested
Campylobacter jejuni in penguins, Antarctica.
Petra Griekspoor et al.
Emerging infectious diseases, 15(5), 847-848 (2009-05-01)
Beatriz del Rio et al.
Clinical and vaccine immunology : CVI, 15(9), 1429-1435 (2008-07-18)
Mucosal immunization is advantageous over other routes of antigen delivery because it can induce both mucosal and systemic immune responses. Our goal was to develop a mucosal delivery vehicle based on bacteria generally regarded as safe, such as Lactobacillus spp.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service