Skip to Content
Merck

78362

Atto 490LS NHS ester

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352108
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥90% (HPLC), ≥90% (degree of coupling)

form

solid

mol wt

Mw 793 g/mol

manufacturer/tradename

ATTO-TEC GmbH

λ

in PBS, pH 7.4

UV absorption

λ: 491-497 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 490LS is a new fluorescent label featuring an extraordinary large Stokes-Shift of 165 nm. Thus the emission spectrum is almost completely separated from its absorption spectrum, making the dye highly suitable for multiplexing experiments, in particular in combination with Atto 488 and Atto 514. Atto 490LS is very hydrophillic and shows excellent water solubility. The dye exhibits a relatively high fluorescence quantum yield, which is only slightly reduced after conjugation to biomolecules, e.g. proteins, even at high degrees of labeling (DOL). Atto 490LS is an anionic dye. After conjugation to a substrate the dye carries a net electrical charge of -1.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library