Skip to Content

77866

Fluorescent IEF-Marker pI 5.5 solution

3 mg/mL in 15 mM HCl

Synonym(s):

IEF-Marker fluorescent pI 5.5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41105336
NACRES:
NA.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

mol wt

Mw 321.27 g/mol

concentration

3 mg/mL in 15 mM HCl

fluorescence

λex 325 nm; λem 412 nm in 0.1 M citrate pH 5.5

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

2-8°C

General description

filtered through a 0.45 μm membrane filter

Application

suitable as IEF-Marker for HPCE and for IEF-gels

Preparation Note

For use in HPCE the marker should be diluted 1:100 in a suitable ampholyte solution (e.g. 2% ampholyte 3-10), UV-detection at 280 nm (20 °C). For use in IEF-gels the solution can be loaded directly onto the gel (max. 1 μl per lane).


ppe

Eyeshields, Gloves, type N95 (US)

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 2

flash_point_f

No data available

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library