Skip to Content
Merck

76559

2-Oleoyl-1-palmitoyl-sn-glycero-3-phospho-rac-(1-glycerol) ammonium salt

≥97.0% (TLC)

Synonym(s):

(9Z)-9-Octadecenoic acid (1R)-1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-2-[(1-oxohexadecyl)oxy]ethyl ester monoammonium salt, 1-Hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol) ammonium salt, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoglycerol ammonium salt, L-α-Phosphatidyl-DL-glycerol, β-oleoyl-γ-palmitoyl ammonium salt, PG(16:0/18:1(9Z)), POPG-NH4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C40H80NO10P
CAS Number:
Molecular Weight:
766.04
NACRES:
NA.85
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

assay

≥97.0% (TLC)

form

solid

solubility

chloroform: 20 mg/mL, clear, colorless

functional group

ester

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

N.CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)OCC(O)CO)OC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C40H77O10P.H3N/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-40(44)50-38(36-49-51(45,46)48-34-37(42)33-41)35-47-39(43)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,37-38,41-42H,3-16,19-36H2,1-2H3,(H,45,46);1H3/b18-17-;/t37?,38-;/m1./s1

InChI key

WFYVYFVKHCDATR-XQYKCTAGSA-N

Application

POPG is widely used in studies as an ingredient in negatively charged synthetic lipid monolayers and bilayers.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library