This item is supplied in solid form as a powder.
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
assay
≥90% (HPLC)
form
powder
storage temp.
2-8°C
SMILES string
[Cl-].OC[C@H]1O[C@@H](Oc2cc3c(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)cc(O)cc3[o+]c2-c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C27H30O16.ClH/c28-7-17-19(33)21(35)23(37)26(42-17)40-15-5-10(30)4-14-11(15)6-16(25(39-14)9-1-2-12(31)13(32)3-9)41-27-24(38)22(36)20(34)18(8-29)43-27;/h1-6,17-24,26-29,33-38H,7-8H2,(H2-,30,31,32);1H/t17-,18-,19-,20-,21+,22+,23-,24-,26-,27-;/m1./s1
InChI key
QDVZZZBBPRFPDG-DHJOXOLYSA-N
Biochem/physiol Actions
Packaging
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥90% (HPLC) | assay ≥95% (HPLC) | assay ≥98.0% (HPLC) | assay ≥90% (HPLC) |
| form powder | form - | form - | form powder |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level 100 |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
This product is liquid or solid?
1 answer-
Helpful?
-


