Skip to Content
Merck
All Photos(1)

Key Documents

71755

Sigma-Aldrich

Sodium nitrate

BioUltra, ≥99.0% (T)

Synonym(s):

Chile salpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.26
Assay:
≥99.0% (T)
form:
solid
solubility:
H2O: 1 M at 20 °C, clear, colorless

product line

BioUltra

Quality Level

Assay

≥99.0% (T)

form

solid

impurities

insoluble matter, passes filter test

pH

5.5-8.0 (25 °C, 1 M in H2O)

mp

306 °C (dec.) (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤5 mg/kg
iodate (IO3-): ≤5 mg/kg
nitrite (NO2-): ≤10 mg/kg
phosphate (PO43-): ≤2 mg/kg
sulfate (SO42-): ≤30 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤20 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 328 nm in H2O at 1 M

SMILES string

[Na+].[O-][N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Sodium nitrate has the ability to reduce acetylcholine receptor clustering, that is stimulated by agrin. It is mainly used by the meat industry to inhibit the development of microbes. It also helps in retaining the colour of conserved and cured meats.

Other Notes

Promotes protoplast fusion; Provides a source of nitrogen in certain culture media

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F. Czapek
Beitr. Chem. Physiol. Pathol., 1, 540-540 (1902)
Sodium nitrate decreases agrin-induced acetylcholine receptor clustering
Jarosz J, et al.
BMC Bioinformatics, 17(1), 20-20 (2016)
A.W. Ruensink
Methods in Enzymology, 58, 539-539 (1979)
Suna Choi et al.
Journal of controlled release : official journal of the Controlled Release Society, 131(1), 70-76 (2008-08-12)
One of the most important requirements for non-viral gene delivery systems is the ability to mediate high levels of gene expression with low toxicity. After the DNA/vector complexes are taken up by cells through endocytosis, DNA is typically contained within
Filip J Larsen et al.
The American journal of clinical nutrition, 99(4), 843-850 (2014-02-07)
Nitrate, which is an inorganic anion abundant in vegetables, increases the efficiency of isolated human mitochondria. Such an effect might be reflected in changes in the resting metabolic rate (RMR) and formation of reactive oxygen species. The bioactivation of nitrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service