Skip to Content
Merck

69025

1-Methylpyrene

suitable for fluorescence, ≥97.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H12
CAS Number:
Molecular Weight:
216.28
UNSPSC Code:
12352108
NACRES:
NA.32
PubChem Substance ID:
EC Number:
219-178-8
Beilstein/REAXYS Number:
1868500
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (GC)

form

solid

solubility

DMSO: soluble, acetonitrile: soluble, chloroform: soluble

fluorescence

λex 340 nm; λem 486 nm in chloroform, λex 346 nm; λem 378 nm in DMSO

suitability

suitable for fluorescence

SMILES string

Cc1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C17H12/c1-11-5-6-14-8-7-12-3-2-4-13-9-10-15(11)17(14)16(12)13/h2-10H,1H3

InChI key

KBSPJIWZDWBDGM-UHFFFAOYSA-N

Analysis Note

In addition to the emission maximum at 378 nm, there are lower maxima at 398, 420 and 445 nm

Other Notes

Fluorescent probe for the determination of micellar aggregation number


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library