Skip to Content
Merck

68599

UV-VIS detectable IEF Marker solution

pI 7.2, 1 mg/mL in 10 mM NaOH, 0.45 μm filtered

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.26
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

0.45 μm filtered

form

liquid

concentration

1 mg/mL in 10 mM NaOH

storage temp.

2-8°C

Application

Suitable as IEF marker for HPCE and IEF gels.


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library