Skip to Content

61786

D-Glyceric acid sodium salt

≥95.0% (TLC)

Synonym(s):

(R)-2,3-Dihydroxypropanoic acid sodium salt, Sodium D-glycerate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H6O4 · xNa+
CAS Number:
Molecular Weight:
106.08 (free acid basis)
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0% (TLC)

form

powder or crystals

optical activity

[α]/D 18.5±3.0°, c = 1 in water

color

white

SMILES string

O[C@H](CO)C(=O)O

InChI

1S/C3H6O4/c4-1-2(5)3(6)7/h2,4-5H,1H2,(H,6,7)/t2-/m1/s1

InChI key

RBNPOMFGQQGHHO-UWTATZPHSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library