Skip to Content

59937

D-myo-Inositol 1-monophosphate dipotassium salt

≥96% (HPLC)

Synonym(s):

D-myo-Inosit 1-phosphate dipotassium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H11K2O9P
CAS Number:
Molecular Weight:
336.32
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥96% (HPLC)

form

solid

color

white

storage temp.

−20°C

SMILES string

[K+].[K+].O[C@H]1[C@H](O)[C@@H](O)[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H]1O

InChI

1S/C6H13O9P.2K/c7-1-2(8)4(10)6(5(11)3(1)9)15-16(12,13)14;;/h1-11H,(H2,12,13,14);;/q;2*+1/p-2/t1-,2-,3+,4-,5-,6-;;/m1../s1

InChI key

HODLUYREWASOIA-PKBSSWBPSA-L

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library