Skip to Content
Merck

Skip To

59304

2-Propanol

greener alternative

≥99.5% (GC), for molecular biology, BioUltra

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352104
EC Number:
200-661-7
MDL number:
Beilstein/REAXYS Number:
635639
Assay:
≥99.5% (GC)
Grade:
Molecular Biology
Technique(s):
DNA extraction: suitable, RNA extraction: suitable
Bp:
82 °C (lit.)
Vapor pressure:
33 mmHg ( 20 °C), 44 mmHg ( 25 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)

grade

Molecular Biology

Quality Segment

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C), 44 mmHg ( 25 °C)

product line

BioUltra

assay

≥99.5% (GC)

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

DNA extraction: suitable, RNA extraction: suitable

impurities

DNases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected

evapn. residue

≤0.001%

color

colorless

refractive index

n20/D 1.377 (lit.), n20/D 1.378

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

solubility

water: soluble (completely)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CC(C)O

cation traces

Al: ≤0.5 mg/kg, Ba: ≤0.1 mg/kg, Bi: ≤0.1 mg/kg, Ca: ≤0.5 mg/kg, Cd: ≤0.05 mg/kg, Co: ≤0.02 mg/kg, Cr: ≤0.02 mg/kg, Cu: ≤0.02 mg/kg, Fe: ≤0.1 mg/kg, K: ≤0.5 mg/kg, Li: ≤0.1 mg/kg, Mg: ≤0.1 mg/kg, Mn: ≤0.02 mg/kg, Mo: ≤0.1 mg/kg, Na: ≤0.5 mg/kg, Ni: ≤0.02 mg/kg, Pb: ≤0.1 mg/kg, Sr: ≤0.1 mg/kg, Zn: ≤0.1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.02, λ: 280 nm Amax: 0.01

suitability

suitable for molecular biology

greener alternative category

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

General description

2-propanol is a clear, colorless, polar organic solvent commonly used in chemical and biochemical laboratories. It will dissolve a wide range of chemicals, and evaporates quickly. Isopropanol is commonly used in molecular biology for DNA and RNA extraction through precipitation. Unlike ethanol, which requires 2.5 times the volume of the nucleic acid-containing sample, isopropanol can be added in an equal volume. This causes precipitation to occur even without cooling, making it advantageous in reducing the overall volume of the sample. Moreover, it has applications in quantifying hydroxyproline levels, assessing cell viability through MTT assays, and contributing to microarray hybridization processes.

Application

2-Propanol has been used:
  • to precipitate genomic DNA extracted from flies
  • to precipitate genomic DNA extracted from xenograft primary tumors and organs
  • to immunoprecipitate 5-bromo-2-deoxyuridine (BrdU)

Biochem/physiol Actions

Isopropanol (2-Propanol) is widely used in molecular biology for the precipitation of DNA during extraction from cells and tissues. BioUltra IPA is suitable for DNA precipitation.

Features and Benefits

  • Suitable as a reagent for Biochemical and Molecular Biology Research
  • BioUltra grade, meets strict purity standards through rigorous testing
  • Free from DNase, NICKase, RNase, and Protease
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This solvent is safe to use and thus aligns with "Safer Solvents and Auxiliaries". Click here for more information.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I951624137-M190764
assay

≥99.5% (GC)

assay

≥99.5%

assay

≥99.5% (GC)

assay

≥99.5%

technique(s)

DNA extraction: suitable, RNA extraction: suitable

technique(s)

DNA extraction: suitable, RNA extraction: suitable

technique(s)

GC/GC: suitable

technique(s)

RNA extraction: suitable

grade

Molecular Biology

grade

Molecular Biology

grade

-

grade

ACS reagent

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

solubility

water: soluble (completely)

solubility

water: soluble (completely)

solubility

-

solubility

H2O: passes test

form

liquid

form

liquid

form

-

form

liquid


Still not finding the right product?

Explore all of our products under 2-Propanol


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

53.6 °F - closed cup

flash_point_c

12.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. is it LCMS grade

    1 answer
    1. This product is molecular biology grade. If you are looking for a LC-MS grade isopropanol, please see product 102781.

      Helpful?

Reviews

No rating value

Active Filters