Skip to Content
Merck
All Photos(1)

Key Documents

56759

Sigma-Aldrich

Atto 425

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27NO6
CAS Number:
Molecular Weight:
401.45
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥90% (HPLC)

form

(Report result)

manufacturer/tradename

ATTO-TEC GmbH

transmittance

254 nm, >90%
425 nm, >90%

fluorescence

λex 440 nm; λem 484 nm

λ

in PBS, pH 7.4
in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 423-429 nm Amax

suitability

suitable for fluorescence

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCOC(=O)C1=Cc2cc3C(C)CC(C)(C)N(CCCC(O)=O)c3cc2OC1=O

InChI

1S/C22H27NO6/c1-5-28-20(26)16-10-14-9-15-13(2)12-22(3,4)23(8-6-7-19(24)25)17(15)11-18(14)29-21(16)27/h9-11,13H,5-8,12H2,1-4H3,(H,24,25)

InChI key

WNDDWSAHNYBXKY-UHFFFAOYSA-N

Application

Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation. Atto 425 is suitable for preparation of fluorescence labeling reagents.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Katrin G Heinze et al.
Biophysical journal, 86(1 Pt 1), 506-516 (2003-12-26)
Confocal fluorescence spectroscopy is a versatile method for studying dynamics and interactions of biomolecules in their native environment with minimal interference with the observed system. Analyzing coincident fluctuations induced by single molecule movement in spectrally distinct detection channels, dual-color fluorescence
Triple FRET: a tool for studying long-range molecular interactions.
Elke Haustein et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 4(7), 745-748 (2003-08-07)
Anna I Sulatskaya et al.
The journal of physical chemistry. B, 116(8), 2538-2544 (2012-01-25)
Benzothiazole dye thioflavin T (ThT) is a sensitive probe for amyloid fibril detection. The ThT probing is based on its unique ability to form highly fluorescent complexes with amyloid and amyloid-like fibrils. In this work we propose an approach of
Frequency domain fluorescence microspectrometry: Application to cellular uptake and drug distribution.
Praus, P., et al.
Spectroscopy: An International Journal, 24, 303-307 (2010)
Nermin Seda Kehr et al.
Nanoscale, 2(4), 601-605 (2010-07-21)
Microcontact printing (mCP) is used to immobilize dyes and peptides asymmetrically, by a "peptide coupling" reaction, on monolayers of zeolite L crystals in the contact area between the stamp and the surface of the monolayer. Chemically patterned surfaces of monolayers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service