Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
description
for research or for further manufacturing use
sterility
sterile
form
liquid
technique(s)
cell culture | mammalian: suitable, cell culture | stem cell: suitable
components
NaHCO3: 350 mg/L
phenol red: no
shipped in
ambient
Application
1 of 1
This Item | |||
|---|---|---|---|
| description HBSS Modified, with calcium, with magnesium, without phenol red, liquid, suitable for cell culture | description HBSS Modified, with phenol red, without calcium, without magnesium, liquid | description Modified, with sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture | description Modified, with sodium bicarbonate, without calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture |
| form liquid | form liquid | form liquid | form liquid |
| technique(s) cell culture | stem cell: suitable, cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable, cell culture | stem cell: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable |
| sterility sterile | sterility sterile | sterility sterile-filtered | sterility sterile-filtered |
| components NaHCO3: 350 mg/L | components NaHCO3: 350 mg/L | components phenol red: no | components NaHCO3: 0.35 g/L |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
| description for research or for further manufacturing use | description for research or for further manufacturing use | description - | description - |
Still not finding the right product?
Explore all of our products under Hanks′ Balanced Salt solution
Storage Class
10 - Combustible liquids
WGK
nwg
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


