Skip to Content
Merck

51797

Atto MB2 maleimide

Methylene Blue derivative, ≥90% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352111
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (HPLC), ≥90% (degree of coupling)

form

solid

manufacturer/tradename

ATTO-TEC GmbH

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 655-661 nm Amax

storage temp.

−20°C

Application

ATTO MB2 is a derivative of the well-known redox dye Methylene Blue. The dye can be reversibly reduced to the colorless leuko form. Upon oxidation the dye recovers and the absorption is fully restored. In common with most ATTO-labels the dye shows a high extinction coefficient. The dye is moderately hydrophilic. ATTO MB2 is a cationic dye. After coupling to a substrate the dye carries a net electrical charge of +1. Maleimides are well suited for coupling to thiol groups. This is similar to iodacetamides, but maleimides react more selectively with thiols. They do not show significant reaction with histidine or methionine. Hydrolysis of maleimides to a mixture of isomeric nonreactive maleamic acids can compete significantly with thiol modification, particularly above pH 8. Maleimides may be used for labeling of amines, which usually requires a higher pH than reaction of maleimides with thiols.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library