Skip to Content
Merck
All Photos(1)

Documents

51416C

SAFC

Minimum Essential Medium

with Earle′s Balanced Salts, with non-essential amino acids, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Minimum Essential Medium Eagle, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

components

Earle’s salts (5% CO2): yes
L-glutamine: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Minimum Essential Medium (MEM), developed by Harry Eagle is one of the most widely used synthetic cell culture media.

Application

Minimum Essential Medium has been used to culture U87 cells (glioblastoma cell line), to study the antitumor action of decane-1,2-diol derivatives. It has also been used to culture SK-Mel-28 (melanoma cell line) cells.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Autophagic flux blockage by accumulation of weakly basic tenovins leads to elimination of B-Raf mutant tumour cells that survive vemurafenib
Ladds MJGW, et al.
PLoS ONE, 13(4), e0195956-e0195956 (2018)
Anisha Viswanathan et al.
Cells, 8(12) (2019-12-18)
Glioblastoma (GB), a grade IV glioma, with high heterogeneity and chemoresistance, obligates a multidimensional antagonist to debilitate its competence. Considering the previous reports on thioesters as antitumor compounds, this paper investigates on use of this densely functionalized sulphur rich molecule
Aaminah Khan et al.
Oncogene, 39(17), 3555-3570 (2020-03-04)
Amplification of the MYCN oncogene occurs in ~25% of primary neuroblastomas and is the single most powerful biological marker of poor prognosis in this disease. MYCN transcriptionally regulates a range of biological processes important for cancer, including cell metabolism. The
Anisha Viswanathan et al.
European journal of pharmacology, 837, 105-116 (2018-09-05)
Glioblastoma remains the most common and aggressive type of malignant brain tumor among adults thus, considerable attention has been given to discovery of novel anti-tumor drugs for its treatment. This study reports the synthesis of a series of twelve novel
Tatsuhiro Tsujimoto et al.
World journal of gastroenterology, 15(44), 5533-5540 (2009-11-26)
To investigate the possible use of the multiple cytokine production modulator, Y-40138, as a novel immunotherapy in the rat nonalcoholic steatohepatitis (NASH) model. We allocated 6-wk-old male F344 rats to choline-supplemented, L-amino acid-defined (CSAA) diet (control group), CSAA diet +

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service