Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352202
NACRES:
NA.77
EC Number:
232-697-4
Biological source:
human
Form:
lyophilized
Technique(s):
cell culture | mammalian: suitable
Product Name
VitroCol®, Collagen from Human neonatal fibroblasts, 97% Type I with the remainder of Type III collagen, suitable for cell culture
biological source
human
sterility
sterile-filtered
form
lyophilized
packaging
pkg of 15 mg
manufacturer/tradename
Advanced BioMatrix
technique(s)
cell culture | mammalian: suitable
impurities
<1 EU/mL Endotoxin level
shipped in
wet ice
storage temp.
−20°C
General description
VitroCol® collagen is naturally secreted from human neo-natal fibroblast cells. The human fibroblasts are cultured in optimal conditions allowing the fibroblasts to naturally and efficiently secret extracellular matrix. The extracellular matrix is then processed and purified to yield the naturally produced human collagen.
VitroCol® is approximately 97% Type I human collagen with the remainder being comprised of Type III collagen. It contains high monomer content as measured by gel permeation chromatography.
VitroCol® is especially ideal for human cell culture systems when coating of surfaces and providing preparations of thin layers of culturing cells. It is not recommend for the formation of a solid gel. VitroCol® is supplied as a sterile, lyophilized powder.
VitroCol® is approximately 97% Type I human collagen with the remainder being comprised of Type III collagen. It contains high monomer content as measured by gel permeation chromatography.
VitroCol® is especially ideal for human cell culture systems when coating of surfaces and providing preparations of thin layers of culturing cells. It is not recommend for the formation of a solid gel. VitroCol® is supplied as a sterile, lyophilized powder.
Preparation Note
This product is supplied as a lyophilized powder with 15 mg of human collagen. When reconstituted with 5 ml of sterile 0.01 N HCl, a concentration of approximately 3 mg/ml is achieved.
Legal Information
VitroCol is a registered trademark of Advanced BioMatrix, Inc.
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.