Skip to Content
Merck
All Photos(1)

Key Documents

47140

Sigma-Aldrich

4-Fluoro-7-nitrobenzofurazan

BioReagent, suitable for fluorescence, ≥98.0% (HPLC)

Synonym(s):

NBD-F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2FN3O3
CAS Number:
Molecular Weight:
183.10
Beilstein:
1077571
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥98.0% (HPLC)

mp

52-54 °C (lit.)

fluorescence

λex 470 nm; λem 528 nm in ethanol
λex 470 nm; λem 550 nm (Amino acid adducts)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(F)c2nonc12

InChI

1S/C6H2FN3O3/c7-3-1-2-4(10(11)12)6-5(3)8-13-9-6/h1-2H

InChI key

PGZIDERTDJHJFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Fluoro-7-nitrobenzofurazan (NBD-F) is used as a protein, amino acid and drug fluorescent labeling reagent in HPLC pre-column derivatization procedures.
Useful for fluorescent labeling of amino acids and amines in HPLC

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Nasr Y Khalil
Talanta, 80(3), 1251-1256 (2009-12-17)
A fully automated, rapid and highly sensitive HPLC method with automated sample pre-treatment by column-switching system and fluorescence detection has been developed for the trace quantitative determination of the new antidepressant reboxetine (RBX) in human plasma. A simple pre-column derivatization
Chiaki Aoyama et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 25(1), 63-65 (2009-01-14)
A simple and sensitive determination method for selenomethionine (Se-Met) using an HPLC-fluorescence detection system coupled with an on-line electrochemical reactor has been developed. NBD-F (4-fluoro-7-nitro-2,1,3-benzoxadiazole) was used as the fluorescent derivatization reagent for Se-Met. NBD-Se-Met was separated from NBD-derivatives of
Yang-Ki Park et al.
Journal of chromatography. A, 1217(20), 3357-3361 (2010-04-13)
Single drop microextraction (SDME) can be in-line coupled with capillary electrophoresis by attaching a drop to the tip of a capillary. With a 2-layer drop comprised of an aqueous basic acceptor phase covered with a thin organic layer, acidic analytes
Yanting Song et al.
Amino acids, 42(5), 1897-1902 (2011-04-21)
A high-performance liquid chromatography method in which fluorescence detection is used for the simultaneous determination of 21 amino acids is proposed. Amino acids were derivatized with 4-fluoro-7-nitro-2,1,3-benzoxadiazole (NBD-F) and then separated on a monolithic silica column (MonoClad C18-HS, 150 mm×3
Hongyi Zhang et al.
Analytical and bioanalytical chemistry, 386(5), 1387-1394 (2006-09-06)
An in-capillary derivatization of amino acids and peptides with 4-fluoro-7-nitro-2,1,3-benzoxadiazole (NBD-F) was developed for their subsequent capillary electrophoretic analysis with laser-induced fluorescence detection (lambda (ex)=488 nm). The in-capillary derivatization was achieved in zone-passing mode by introducing successive plugs of sample

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service