Skip to Content
Merck
All Photos(1)

Documents

46985

Sigma-Aldrich

Fluorescein-5-thiosemicarbazide

suitable for fluorescence, ~80% (HPCE)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H15N3O5S
CAS Number:
Molecular Weight:
421.43
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.32

Assay

~80% (HPCE)

form

solid

solubility

DMF: soluble
H2O: soluble

fluorescence

λex 492 nm; λem 516 nm in 0.1 M Tris pH 9.0

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

NNC(=S)Nc1ccc(c(c1)C(O)=O)C2=C3C=CC(=O)C=C3Oc4cc(O)ccc24

InChI

1S/C21H15N3O5S/c22-24-21(30)23-10-1-4-13(16(7-10)20(27)28)19-14-5-2-11(25)8-17(14)29-18-9-12(26)3-6-15(18)19/h1-9,25H,22H2,(H,27,28)(H2,23,24,30)

InChI key

MEUCHQDLZYLNQY-UHFFFAOYSA-N

General description

Fluorescein-5-thiosemicarbazide, also known as FTC, is a fluorescent probe. The FTSC fluorescent spectra is like FITC, a preferred fluorescent tag for proteomics. The fluorescence spectral properties of FTSC-labeled vesicles obtained by reductive amination are fully compatible with the argon laser of fluorescent instruments.

Application

Fluorescein-5-thiosemicarbazide can be used as a fluorescent labeling reagent to label:
  • Cell-surface functional groups (glycophorins) in the study of the effect of deoxygenation in the red blood cell membrane.
  • Saccharides applicable in polysaccharide imaging in live cells.
  • Chondroitin sulfate nanogels applicable in cell-specific drug delivery applications.

Features and Benefits

Fluorescein-5-thiosemicarbazide has the following benefits -

  • It displays strong fluorescence property.
  • Shows an intrinsic reactivity of the thiosemicarbazide group towards the aldehyde group.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D Corbett et al.
Biophysical journal, 66(1), 25-30 (1994-01-01)
We have used the fluorescence photobleaching recovery technique to study the dependence on oxygen tension of the lateral mobility of fluorescently labeled band 3, the phospholipid analogue fluorescein phosphatidylethanolamine, and glycophorins in normal red blood cell membranes. Band 3 protein
Hua Tian et al.
Journal of cellular and molecular medicine, 21(1), 107-120 (2016-08-12)
Oxidative stress and inflammation play important roles in the pathogenesis of cardiovascular disease (CVD). Oxidative stress-induced desialylation is considered to be a primary step in atherogenic modification, and therefore, the attenuation of oxidative stress and/or inflammatory reactions may ameliorate CVD.
Deoxygenation affects fluorescence photobleaching recovery measurements of red cell membrane protein lateral mobility
Corbett JD, et al.
Biophysical Journal, 66(1), 25-30 (1994)
Injectable hydrogel-incorporated cancer cell-specific cisplatin releasing nanogels for targeted drug delivery
Gil MS, et al.
Journal of Material Chemistry B: Materials for Biology and Medicine, 5(34), 7140-7152 (2017)
Yun Bai et al.
Nucleic acids research, 41(16), 7861-7874 (2013-06-21)
The 3' untranslated region (3'UTR) of hepatitis C virus (HCV) messenger RNA stimulates viral translation by an undetermined mechanism. We identified a high affinity interaction, conserved among different HCV genotypes, between the HCV 3'UTR and the host ribosome. The 3'UTR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service