Skip to Content
Merck

45714

Eriocitrin

≥98.0% (HPLC)

Synonym(s):

(S)-3′,4′,5,7-Tetrahydroxyflavanone-7-[6-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], Eriodictioside, Eriodictyol 7-O-rutinoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H32O15
CAS Number:
Molecular Weight:
596.53
UNSPSC Code:
12352205
NACRES:
NA.47
PubChem Substance ID:
EC Number:
236-668-7
Beilstein/REAXYS Number:
1304401
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98.0% (HPLC)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](Oc3cc(O)c4C(=O)C[C@H](Oc4c3)c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H32O15/c1-9-20(32)22(34)24(36)26(39-9)38-8-18-21(33)23(35)25(37)27(42-18)40-11-5-14(30)19-15(31)7-16(41-17(19)6-11)10-2-3-12(28)13(29)4-10/h2-6,9,16,18,20-30,32-37H,7-8H2,1H3/t9-,16-,18+,20-,21+,22+,23-,24+,25+,26+,27+/m0/s1

InChI key

OMQADRGFMLGFJF-MNPJBKLOSA-N

Biochem/physiol Actions

Flavone glycoside found in lemon. Administration prior to acute exercise helped to maintain the redox status of liver glutathione. Without eriocitrin, it shifts markedly toward oxidized form.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Still not finding the right product?

Explore all of our products under Eriocitrin


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library