Skip to Content
Merck

45703

Erythromycin standard solution

1 mg/mL in H2O

Synonym(s):

Erythromycin from Streptomyces erythreus

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C37H67NO13
CAS Number:
Molecular Weight:
733.93
UNSPSC Code:
51282304
NACRES:
NA.85
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
8183758
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Streptomyces erythreus (strains)

agency

EPA 1694

form

liquid

concentration

1 mg/mL in H2O

color

colorless

antibiotic activity spectrum

Gram-positive bacteria

application(s)

environmental

mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CC[C@H]1OC(=O)[C@H](C)C(O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)C(C)C(OC3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)C(O)[C@]1(C)O

InChI

1S/C37H67NO13/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1

InChI key

ULGZDMOVFRHVEP-RWJQBGPGSA-N

General description

Chemical structure: macrolide

Application

Erythromycin is an antibiotic produced by growth of certain strains of Streptomyces erythreus. This product is composed largely of erythromycin A with small amounts of erythromycins B and C and is recommended for concentration at 100 mg/L. Concentrations between 50 and 200 mg/L have also proven effective in controlling bacterial growth. Erythromycin has been used as a motilin receptor agonist, to block respiratory glycoconjugate secretion in human airways in vitro, and for selecting plasmid-cured and recombinant lactococcus lactis MG1363 strains. .

Biochem/physiol Actions

Mode of Action: Erythromycin acts by inhibiting elongation at the transpeptidation step, specifically aminoacyl translocation from the A-site to P-site by binding to the 50s subunit of the bacterial 70s rRNA complex.

Antimicrobial Spectrum: This product acts against both gram-negative and gram-positive bacteria.

Packaging

10 ml

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Store under inert gas.

Disclaimer

This Erythromycin solution is concentrated at 1 mg/mL in H2O and is stable in solution at 37°C for 3 days. Stock solutions should be stored at 2-8°C.


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library