Skip to Content
Merck

44714

(S)-Mevalonic acid lithium salt

≥96.0% (GC)

Synonym(s):

Lithium (S)-3,5-dihydroxy-3-methyl-pentanoate, Lithium (S)-3,5-dihydroxy-3-methyl-valerate, S-MVA-Li

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H12O4 · xLi+
CAS Number:
Molecular Weight:
148.16 (free acid basis)
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.25
Assay:
≥96.0% (GC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥96.0% (GC)

optical activity

[α]/D +25.0±3.0°, c = 0.1 in 1 M HCl

storage temp.

2-8°C

SMILES string

O[C@@](CCO)(CC(=O)O)C

InChI

1S/C6H12O4/c1-6(10,2-3-7)4-5(8)9/h7,10H,2-4H2,1H3,(H,8,9)/t6-/m0/s1

InChI key

KJTLQQUUPVSXIM-LURJTMIESA-N

Biochem/physiol Actions

Metabolite of the mevalonate pathway, which plays a key role in the biosynthesis of sterols, dolichol, heme and ubiquinone. Of interest for research in the disease areas oncology, autoimmune diseases, artherosclerosis and Alzheimer disease, as well as for inherited deficiencies of mevalonate kinase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Still not finding the right product?

Explore all of our products under (S)-Mevalonic acid lithium salt


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library