Skip to Content

44611

Durcupan ACM

single component A, M epoxy resin

Synonym(s):

ACM resins

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
13111001
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

impurities

bisphenol A epoxy resin

General description

Awater-soluble embedding medium for EM based on an aliphatic polyepoxide.

Application

DurcupanACM, single component A, M epoxy resin has been used for:
  • A method to prepare large resin sections for counting myelinated axons in rodent CNS and PNS structures.: This study presents a technique using Durcupan ACM for preparing large resin sections, enhancing the analysis of myelinated axons in rodent central and peripheral nervous systems.
  • Three-dimensional immersive virtual reality for studying cellular compartments in 3D models from EM preparations of neural tissues.: This research utilizes Durcupan ACM in electron microscopy to create 3D models of neural tissues, facilitating studies via immersive virtual reality.
  • Long-term effect of prazosin and losartan administration on blood pressure, heart, carotid artery, and acetylcholine induced dilation of cardiovascular system of young Wistar rats and SHR.: Explores the cardiovascular effects in rats using Durcupan ACM embedded samples for detailed tissue analysis.
  • Embedding material for electron microscopy on the basis of Araldite®.

Features and Benefits

Mixtures of Durcupan ACM present an interesting alternative to methacrylates for embedding sections for electron microscopy. They present the advantage that they harden uniformly, practically without shrinkage. All fixing agents normally used for electronmicroscopic work are also suitable for the Durcupan ACM embedding process. This process consists of three phases: dehydration of the tissue with acetone or ethyl alcohol, infiltration with the embedding agent, followed by hardening.

Legal Information

Araldite is a registered trademark of Huntsman Advanced Materials Inc.
Durcupan is a trademark of Sigma-Aldrich Chemie GmbH


Still not finding the right product?

Explore all of our products under Durcupan ACM


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

377.6 °F

flash_point_c

192 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library