Skip to Content
Merck
All Photos(1)

Documents

404A-1

Sigma-Aldrich

C4d Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (404A-14)
vial of 0.5 mL concentrate (404A-15)
bottle of 1.0 mL predilute (404A-17)
vial of 1.0 mL concentrate (404A-16)
bottle of 7.0 mL predilute (404A-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

control

lymph node, tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, membranous

Gene Information

human ... C4B(721)

General description

C4d is a stable split product remnant of classical complement activation which becomes covalently bound to endothelium and basement membrane, after induction of the classical antibody-induced pathway. As an established marker of antibody-mediated acute renal allograft rejection and its proclivity for endothelium, this component can be detected in peritubular capillaries in both chronic renal allograft rejection as well as hyperacute rejection, acute vascular rejection, acute cellular rejection, and borderline rejection. It has been shown to be a significant predictor of transplant kidney graft survival and is an aid in treating acute rejection.

Quality


IVD

IVD

IVD

RUO

Linkage

C4d Positive Control Slides, Product No. 404S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tian Ye et al.
Lupus science & medicine, 10(2) (2023-12-29)
This study aimed to investigate the clinical significance of exostosin 1 (EXT1) in confirmed and suspected lupus membranous nephropathy (LMN). EXT1 was detected in 67 renal tissues of M-type phospholipase A2 receptor (PLA2R)-negative and ANA-positive membranous nephropathy by immunohistochemistry, and
Chen Jianghua et al.
Clinical transplantation, 19(6), 785-791 (2005-11-30)
To determine the diagnostic and clinical significance of C4d accumulation in renal allografts followed by acute rejection. A total of 158 graft biopsies performed from December 1997 to December 2002 were classified, according to the Banff-97 criteria, into hyperacute rejection
Dorothée Bouron-Dal Soglio et al.
Human pathology, 39(7), 1103-1110 (2008-06-24)
C4d is a marker of the activated complement cascade used to assess the humoral component of rejection, mostly in kidney allograft transplants. The role of C4d deposition has recently been addressed in hepatic allograft but has never been tested in
Gyongyi M Nadasdy et al.
Human pathology, 36(11), 1178-1185 (2005-11-02)
Detection of peritubular capillary (PTC) C4d deposition in tissue sections of renal allograft biopsies became an important aid in the diagnosis of antibody-mediated rejection. Pathologists in many major transplant centers now routinely stain renal allograft biopsies for C4d. Currently, there
Christian A Seemayer et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 22(2), 568-576 (2006-12-14)
Detection of C4d along peritubular capillaries (PTC) in renal allograft biopsies is an independent prognostic marker of poor long-term graft survival. It is typically associated with circulating donor-specific antibodies. Since only little information is available on the best technique to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service