Skip to Content
Merck
All Photos(2)

Key Documents

3952016

Sigma-Aldrich

Schiff′s Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.47
grade:
for analytical purposes
form:
solution

grade

for analytical purposes

form

solution

shelf life

Expiry date on the label

IVD

for in vitro diagnostic use

pH

<2.0

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Application

Intended for use in Sigma Periodic Acid Staining Procedure, 395. Kit is intended for use with blood, bone marrow, tissue touch preparations or routine tissue sections.

Components

Pararosaniline, 1% and sodium metabisulfate, 4%, in hydrochloric acid, 0.25 mol/L

Other Notes

Use Sigma reagent S5133 for staining glycoproteins in polyacrylamide gels.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Met. Corr. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naser Mobarra et al.
International journal of hematology-oncology and stem cell research, 8(4), 20-29 (2015-03-17)
The use of stem cells is considered as an appropriate source in cell therapy and tissue engineering. Differentiation of human induced Pluripotent Stem Cells (hiPSCs) to Hepatocyte-like Cells (HLCs) on mouse embryonic fibroblasts (MEFs) feeders is confronted with several problems
Yan-Yan Song et al.
Oncology letters, 8(1), 431-437 (2014-06-25)
Vasculogenic mimicry (VM) formation is important for invasion and metastasis of tumor cells in gastric adenocarcinoma (GAC). The present study aimed to investigate the association between signal transducer and activator of transcription-3 (STAT3), phosphor-STAT3 (p-STAT3), hypoxia-inducible factor-1α (HIF-1α) and VM
Rüdiger Braun et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 26(13), 3468-3480 (2020-04-08)
The standard treatment of patients with locally advanced rectal cancer consists of preoperative chemoradiotherapy (CRT) followed by surgery. However, the response of individual tumors to CRT is extremely diverse, presenting a clinical dilemma. This broad variability in treatment response is
Priti Chougule et al.
Scandinavian journal of gastroenterology, 47(11), 1334-1343 (2012-09-05)
Cell culture studies of enterocytes are important in many fields. However, there are difficulties in obtaining cell lines from adult human intestine, such as microbial contamination of cultures from the tissue samples, short life span of enterocytes, overgrowth of mesenchymal
Jin-Yong Chung et al.
Environmental health perspectives, 119(11), 1569-1574 (2011-07-09)
Benzo[a]pyrene (B[a]P), a polycyclic aromatic hydrocarbon (PAH), is a ubiquitous environmental pollutant that is currently suspected of being an endocrine disruptor. The testis is an important target for PAHs, yet insufficient attention has been paid to their effects on steroidogenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service