Skip to Content
Merck

38153

8-Hydroxyquinoline-β-D-glucuronide sodium salt

≥98.0% (HPLC)

Synonym(s):

8-Hydroxyquinoline-β-D-glucuronic acid sodium salt, 8-Quinolyl-β-D-glucopyranosiduronic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H14NNaO7
CAS Number:
Molecular Weight:
343.26
NACRES:
NA.83
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98.0% (HPLC)

form

powder

optical activity

[α]/D -84.0±4.0°, c = 1 in H2O

impurities

≤12% water

storage temp.

−20°C

SMILES string

[Na+].O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C([O-])=O)Oc2cccc3cccnc23

InChI

1S/C15H15NO7.Na/c17-10-11(18)13(14(20)21)23-15(12(10)19)22-8-5-1-3-7-4-2-6-16-9(7)8;/h1-6,10-13,15,17-19H,(H,20,21);/q;+1/p-1/t10-,11-,12+,13-,15+;/m0./s1

InChI key

QYRMLNFJXNJXLO-KSOKONAESA-M

Application

Substrate for detection of β-D-glucuronidase, yields a brown precipitate upon cleavage; E. coli detection

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



R D Reinders et al.
Letters in applied microbiology, 30(5), 411-414 (2000-05-03)
8-hydroxyquinoline-beta-D-glucuronide (HQG) was used to improve the presumptive identification of Shiga toxin-producing Escherichia coli O157 (STEC O157) on sorbitol MacConkey agars (SMAC). Advantages of HQG are (i) that it is less expensive than 5-bromo-4-chloro-3-indoxyl-glucuronide; (ii) that it is visible in
A L James et al.
Zentralblatt fur Bakteriologie, Mikrobiologie, und Hygiene. Series A, Medical microbiology, infectious diseases, virology, parasitology, 267(3), 316-321 (1988-01-01)
The use of 8-hydroxyquinoline-beta-D-glucuronide for the demonstration of beta-glucuronidase activity within the family Enterobacteriaceae has been investigated. The compound has been shown to be an effective substrate for the bacterial enzyme and the Michaelis constant for the association has been



Global Trade Item Number

SKUGTIN
38153-100MG04061833471746
38153-500MG04061831158342