Skip to Content
Merck

37161

1,2-Dihexadecyl-sn-glycero-3-phosphoethanolamine

≥99.0% (TLC)

Synonym(s):

PE(O-16:0/O-16:0), 3-sn-Phosphatidylethanolamine, 1,2-dihexadecyl, L-β,γ-Dihexadecyl-α-cephalin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C37H78NO6P
CAS Number:
Molecular Weight:
663.99
UNSPSC Code:
51191904
NACRES:
NA.25
PubChem Substance ID:
EC Number:
262-782-1
Beilstein/REAXYS Number:
2029302
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0% (TLC)

form

powder

functional group

amine

lipid type

phosphoglycerides

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCOC[C@H](COP(O)(=O)OCCN)OCCCCCCCCCCCCCCCC

InChI

1S/C37H78NO6P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-32-41-35-37(36-44-45(39,40)43-34-31-38)42-33-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37H,3-36,38H2,1-2H3,(H,39,40)/t37-/m1/s1

InChI key

UXDBPOWEWOXJCE-DIPNUNPCSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library