Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
culture supernatant
antibody product type
primary antibodies
clone
MRQ-60, monoclonal
description
For In Vitro Diagnostic Use in Select Regions (See Chart)
form
buffered aqueous solution
species reactivity
human
packaging
vial of 0.1 mL concentrate (352M-94), vial of 0.5 mL concentrate (352M-95), bottle of 1.0 mL predilute (352M-97), vial of 1.0 mL concentrate (352M-96), bottle of 7.0 mL predilute (352M-98)
manufacturer/tradename
Cell Marque®
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500
isotype
IgG1κ
control
lung adenocarcinoma, renal cell carcinoma
shipped in
wet ice
storage temp.
2-8°C
visualization
cytoplasmic
Gene Information
human ... NAPSA(9476)
General description
Physical form
Preparation Note
Analysis Note
![]() IVD | ![]() IVD | ![]() IVD | ![]() RUO |
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| clone MRQ-60, monoclonal | clone polyclonal | clone MRQ-50, monoclonal | clone MRQ-3, monoclonal |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form culture supernatant | antibody form Ig fraction of antiserum | antibody form culture supernatant | antibody form culture supernatant |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| biological source mouse | biological source rabbit | biological source mouse | biological source mouse |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.







