Skip to Content
Merck

Skip To

285D

Fetal Hemoglobin Kit

Synonym(s):

Modified Kleihauer-Betke procedure

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
41116121
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 200 manual assays

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

dilution

(for the determination of fetal hemaglobin in blood smears)

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Application

Intended for the acid elution of adult hemoglobin and the semi-quantitative determination of fetal hemaglobin in blood smears.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
285CHT25AHT100A
application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

500

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

-

dilution

(for the determination of fetal hemaglobin in blood smears)

dilution

(for the determination of fetal hemaglobin in blood smears)

dilution

(for histology)

dilution

(for histology)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use


Still not finding the right product?

Explore all of our products under Fetal Hemoglobin Kit


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Citrate Phosphate Buffer Concentrate 2 x 100
    SDS

  • Acid Hematoxylin Solution 2 x 100
    SDS

  • Eosin B Solution, 0.1% aqueous solution 2 x 100
    SDS

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Are slides included in the 285C or 285D Fetal Hemoglobin kits?

    1 answer
    1. The kit has never contained slides. The only kit components in the kit are solutions 2851, 2852, and 2853. The user needs to provide a positive control, such as blood from a newborn or a woman in her third trimester of pregnancy.

      Helpful?

Reviews

No rating value

Active Filters