Skip to Content
Merck
All Photos(1)

Key Documents

21906

Sigma-Aldrich

Carboxymethyl-β-cyclodextrin sodium salt

Synonym(s):

Sodium CM-β-Cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

form

solid

impurities

10% water

mp

~245 °C (dec.)

solubility

H2O: 50 mg/mL, clear to slightly hazy, colorless

SMILES string

O[C@H]1[C@H](O)[C@@H]2O[C@H]3O[C@H](COCC(O)=O)[C@@H](O[C@H]4O[C@H](COCC(O)=O)[C@@H](O[C@H]5O[C@H](COCC(O)=O)[C@@H](O[C@H]6O[C@H](COCC(O)=O)[C@@H](O[C@H]7O[C@H](COCC(O)=O)[C@@H](O[C@H]8O[C@H](COCC(O)=O)[C@@H](O[C@H]1O[C@@H]2COCC(O)=O)[C@@H](O)[C@@H]8O)[C@@H](O)[C@@H]7O)[C@@H](O)[C@@H]6O)[C@@H](O)[C@@H]5O)[C@@H](O)[C@@H]4O)[C@@H](O)[C@@H]3O

InChI

1S/C56H84O49/c57-22(58)8-85-1-15-43-29(71)36(78)50(92-15)100-44-16(2-86-9-23(59)60)94-52(38(80)31(44)73)102-46-18(4-88-11-25(63)64)96-54(40(82)33(46)75)104-48-20(6-90-13-27(67)68)98-56(42(84)35(48)77)105-49-21(7-91-14-28(69)70)97-55(41(83)34(49)76)103-47-19(5-89-12-26(65)66)95-53(39(81)32(47)74)101-45-17(3-87-10-24(61)62)93-51(99-43)37(79)30(45)72/h15-21,29-56,71-84H,1-14H2,(H,57,58)(H,59,60)(H,61,62)(H,63,64)(H,65,66)(H,67,68)(H,69,70)/t15-,16-,17-,18-,19-,20-,21-,29+,30+,31+,32+,33+,34+,35+,36+,37+,38+,39+,40+,41+,42+,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,55-,56-/m1/s1

InChI key

WROHVVIPQXODQM-LYSKQWNXSA-N

General description

Carboxymethyl-β-cyclodextrin, a negatively charged derivative of β-cyclodextrin, is generally used in the chiral analysis of neutral and basic analytes on account of its ability to exhibit counter-current mobility.

Application

Cyclodextrins (CD) are cyclic oligosaccharides composed of D-glucose units connected via α(1→4) glycosidic bonds. Common cyclodextrins contain six (α-CD); seven (β-CD) or eight (gamma-CD) D-glucose units. Cyclodextrins are sometimes derivitized through esterification at positions two, three and/or six.
Carboxymethyl-β-cyclodextrin (CMBCD) is used in the development of recognition and separation technologies to resolve enantiomers based on chiral properties and size. Carboxymethyl-β-cyclodextrin is used in chiral selection and separation by capillary electrophoresis. Carboxymethyl-β-cyclodextrin is used in the development of drug delivery vehicles such as nanocarriers and as a nucleic acid transfection agent.

Analysis Note

average degree of substitution, DS ~3

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral Separations: Methods and Protocols
Methods in Molecular Biology (2004)
Felipe A Oyarzun-Ampuero et al.
Macromolecular bioscience, 12(2), 176-183 (2011-11-24)
A new nanomedicine consisting of chitosan/carboxymethyl-β-cyclodextrin loaded with unfractioned or low-molecular-weight heparin is described and its potential in asthma treatment is evaluated. The nanoparticles are prepared by ionotropic gelation showing a size that between 221 and 729 nm with a
Li-Li Ren et al.
Journal of nanoscience and nanotechnology, 10(11), 7262-7265 (2010-12-09)
Mammalian cochlear hair cells don't regenerate naturally after injury, which usually leave permanent hearing loss. Math1 gene is a positive regulator of hair cell differentiation during cochlear development and was proved to be very critical in hair cell regeneration in
Selective recognition and separation of nucleosides using carboxymethyl-β-cyclodextrin functionalized hybrid magnetic nanoparticles.
Badruddoza AZ, Junwen L, et al.
Colloids and Surfaces, B: Biointerfaces, 92, 223-231 (2012)
Thitirat Mantim et al.
Electrophoresis, 33(2), 388-394 (2011-12-16)
CE methods with capacitively coupled contactless conductivity detection (C(4)D) were developed for the enantiomeric separation of the following stimulants: amphetamine (AP), methamphetamine (MA), ephedrine (EP), pseudoephedrine (PE), norephedrine (NE) and norpseudoephedrine (NPE). Acetic acid (pH 2.5 and 2.8) was found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service