Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.81
UNSPSC Code:
12352202
Form:
powder
Biological source:
bovine
biological source
bovine
Quality Level
form
powder
technique(s)
tissue processing: suitable
storage temp.
−20°C
Application
Selectively soluble components of liver extracts were used in early nutritional studies of essential micronutrients.
Other Notes
Commercially dried liver extract containing B-factors.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A liver concentrate as a source of unrecognized vitamins required by the chick
Richardson, L.R., et al.
The Journal of Nutrition, 24, 65-72 (1942)
Adrienne E Campbell-Washburn et al.
Journal of magnetic resonance imaging : JMRI, 55(6), 1855-1863 (2021-10-21)
MRI T2* and R2* mapping have gained clinical acceptance for noninvasive assessment of iron overload. Lower field MRI may offer increased measurement dynamic range in patients with high iron concentration and may potentially increase MRI accessibility, but it is compromised
Adolfo Cruz-Reséndiz et al.
BMC biotechnology, 20(1), 1-1 (2020-01-22)
The use of biomaterials has been expanded to improve the characteristics of vaccines. Recently we have identified that the peptide PH(1-110) from polyhedrin self-aggregates and incorporates foreign proteins to form particles. We have proposed that this peptide can be used
Global Trade Item Number
| SKU | GTIN |
|---|---|
| F14335-5G | 04061833611951 |
| F14335-10G | 04061836830892 |
| 2023-500G | 04061833627037 |
| 2023-50G | 04061838764690 |
| 2023-100G | 04061838764683 |