Skip to Content
Merck
All Photos(3)

Key Documents

14300

Sigma-Aldrich

Betaine monohydrate

≥99.0% (NT)

Synonym(s):

1-carboxy-n,n,n-trimethylmethanaminium hydroxide, 2-(trimethylammonio)acetate hydrate, glycine betaine hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N+CH2COO- · H2O
CAS Number:
Molecular Weight:
135.16
Beilstein:
3626258
EC Number:
MDL number:
UNSPSC Code:
51171529
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99.0% (NT)

form

solid

solubility

H2O: 0.1 g/mL, clear, colorless

storage temp.

2-8°C

SMILES string

O.C[N+](C)(C)CC([O-])=O

InChI

1S/C5H11NO2.H2O/c1-6(2,3)4-5(7)8;/h4H2,1-3H3;1H2

InChI key

NJZRLXNBGZBREL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Betaine monohydrate:

  • is suitable for use to improve the PCR amplification of GC-rich DNA sequences by reducing the formation of secondary structure caused by GC-rich regions
  • is suitable for use as a component of PCR reaction mixture
  • is suitable for use in a study to identify yield enhancers of PCR
  • has been used to study nitrogen balance and nutrient digestibility in piglets fed diets deficient in methionine and low in compatible osmolytes
  • has also been used to compare the effects of pressure on three layered hydrates

Biochem/physiol Actions

End-product of oxidative metabolism of choline, betaine is a general methyl donor, in particular in a minor pathway of methionine biosynthesis. It is used to treat homocystinuria, which is a defect in the major pathway of methionine biosynthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kovárová et al.
Nucleic acids research, 28(13), E70-E70 (2000-06-28)
Tetramethylammonium (TMA) chloride, dimethyl sulfoxide and formamide are known to increase, under certain conditions, the specificity and efficiency of the polymerase chain reaction (PCR). We compared the ability of several TMA derivatives and some other reagents to increase the specificity
Russell D L Johnstone et al.
Acta crystallographica. Section B, Structural science, 65(Pt 6), 731-748 (2009-11-20)
We report the effect of pressure on the crystal structures of betaine monohydrate (BTM), L-cysteic acid monohydrate (CAM) and S-4-sulfo-L-phenylalanine monohydrate (SPM). All three structures are composed of layers of zwitterionic molecules separated by layers of water molecules. In BTM
Theodore Spilker et al.
Journal of clinical microbiology, 50(9), 3010-3015 (2012-07-13)
The genus Achromobacter currently is comprised of seven species, including Achromobacter xylosoxidans, an opportunistic and nosocomial pathogen that displays broad-spectrum antimicrobial resistance and is recognized as causing chronic respiratory tract infection in persons with cystic fibrosis (CF). To enable strain
Meike Eklund et al.
Archives of animal nutrition, 60(4), 289-300 (2006-08-23)
A balance experiment was carried out to investigate the effects of betaine monohydrate (BET) or betaine derived from condensed molasses solubles (CMS) as a substitute for methionine and choline on nitrogen (N) balance and total tract nutrient digestibility in weaned
Cletus Nunes et al.
Journal of pharmaceutical sciences, 93(1), 38-47 (2003-12-04)
The objectives were (1). to perform solid-state characterization of anhydrous betaine (A) and betaine monohydrate (M), (2). to develop a pressure differential scanning calorimetric (DSC) technique for the quantification of M when present as a minor component in a mixture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service