Skip to Content
Merck
All Photos(1)

Key Documents

137257

SAFC

MES sodium salt

EMPROVE® EVOLVE

Pharma Manufacturing

Synonym(s):

MES, Sodium Salt, 2-(N-Morpholino)ethanesulfonic acid sodium salt, 4-Morpholineethanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12NNaO4S
CAS Number:
Molecular Weight:
217.22
Beilstein:
3765682
MDL number:
UNSPSC Code:
12161700
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin and microbial, tested

pH

5.5-6.7

useful pH range

5.5-6.7

pKa 

6.1
6.1

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

storage temp.

2-30°C

SMILES string

[Na+].[O-]S(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.Na/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);/q;+1/p-1

InChI key

IRHWMYKYLWNHTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

MES Sodium is a biological buffer often referred to as a “Good′s” buffer. The pKa of MES is 5.96 which makes MES an ideal candidate for cell culture media and protein based buffer formulations to maintain a stable environment in solution. MES Sodium is considered to be non-toxic to culture cell lines, highly water soluble and provides high-solution clarity.MES Sodium is used in cell culture media, biopharmaceutical buffer formulations both upstream and downstream and diagnostic reagents. MES based buffers are used in purification bioprocesses of antibodies, peptides, proteins and blood components.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service