Skip to Content
Merck

Skip To

125-50

Collagen Coating Solution (50 ML)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.75
UNSPSC Code:
12352207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

shelf life

1 yr

shipped in

wet ice

storage temp.

2-8°C

General description

Collagen is a major fibrous, structural protein in animals. It is a key component of the extracellular matrix (ECM) and is predominantly present in the connective tissue, ligaments, tendons, and cornea. Collagens consist of three parallel left-handed polypeptide chains coiled to form a right-handed triple helix. They also have a globular domain.

Application

The coating concentration is 1 ml per 10 cm2 surface area of the cell culture ware: 2.5 ml for coating a T-25 flask or a 60 mm tissue culture dish. 7.5 ml for coating a T-75 flask or a 100 mm tissue culture dish.

Add Attachment Factor Solution to the flask and rock the flask gently to distribute the solution evenly to cover the whole culture surface. Coat the culture ware surface for 30 minutes at 37°C or 2 hours or overnight at room temperature. Remove Attachment Factor Solution by aspiration in a sterile hood. The coated flask can be used immediately or store at 4°C for up to 2 weeks.

Biochem/physiol Actions

Collagen is a basic unit for the growth and support of various cells and tissues in the body. Collagen may be involved in wound healing. Collagen might promote cell adhesion and proliferation when used as a coating on iron-manganese alloys (Fe-30%Mn) scaffolds in orthopedic applications.


Storage Class

12 - Non Combustible Liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

Active Filters

  1. 1 Ratings-only review