Skip to Content
Merck
All Photos(1)

Key Documents

114R-1

Sigma-Aldrich

CD14 (EPR3653) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EPR3653, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (114R-14)
vial of 0.5 mL concentrate (114R-15)
bottle of 1.0 mL predilute (114R-17)
vial of 1.0 mL concentrate (114R-16)
bottle of 7.0 mL predilute (114R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, membranous

Gene Information

human ... CD14(929)

General description

CD14 is a surface protein preferentially expressed on monocytes/macrophages. It binds lipopolysaccharide binding protein and recently has been shown to bind apoptotic cells. Anti-CD14 antibody labels Kupffer cells in liver sinusoids. In lymphoid tissues, dendritic cells are distinctly stained. Most other normal tissues are negative. This antibody labels monocyte-macrophages and Langerhans cells in Langerhans cell histiocytosis. Tumor cells are positive in monocytic leukemia and true histiocytic lymphomas for CD14. Sinusoidal histiocytes express CD14 and CD 169, whereas most of the other monocyte-derived cells in reactive lymph node lack these markers. Anti-CD14 labels numerous diffuse large B-cell lymphomas and splenic marginal zone lymphoma but not in other B-cell lymphomas.

Quality


IVD

IVD

IVD

RUO

Linkage

CD14 Positive Control Slides, Product No. 114S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A T Larregina et al.
Nature immunology, 2(12), 1151-1158 (2001-11-10)
Epidermal Langerhans cells (LCs) show extraordinary immunostimulatory capacity and play a key role in the initiation and regulation of immune responses. Studies of LC biology are currently the focus of efforts to engineer immune responses and to better understand the
B Steiniger et al.
Immunology, 92(2), 307-316 (1998-01-01)
The microanatomical structure of human and rat splenic white pulp is compared, with special emphasis on the localization of the marginal zone occupied by immunoglobulin M (IgM)+ IgD-/dull B lymphocytes and its specialized macrophages. Our study reveals that in contrast
H W Ziegler-Heitbrock et al.
Immunology today, 14(3), 121-125 (1993-03-01)
In the past, CD14 has been viewed simply as a useful marker molecule for monocytes and macrophages. Now, new findings on its role in binding of LPS-LBP complexes and in signal transduction have engendered renewed interest in the properties of
Béatrice Marmey et al.
Human pathology, 37(1), 68-77 (2005-12-20)
The mononuclear phagocyte system of human lymphoid tissue comprises macrophages and dendritic cells (DCs). The heterogeneity of the non-DC mononuclear phagocyte population in human lymphoid tissue has been little addressed. Here, we studied the expression of 2 monocyte-derived markers, CD14
C D Gregory et al.
Apoptosis : an international journal on programmed cell death, 4(1), 11-20 (2003-11-25)
In addition to its role as a mediator of innate pro-inflammatory responses following bacterial lipopolysaccharide (LPS) binding, the 55kDa glycosyl-phosphatidylinositol-linked macrophage plasma membrane glycoprotein CD14 is now also known to play a role in phagocytic clearance of apoptotic cells. Although

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service